siRNA Details
| Gene Symbol: 2900073C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013770.1 | - | 1 | 302 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712737.1 | - | 1 | 304 | Chr. 4 - 43748026 - 43748644 [-] on Build GRCm38 | |

2900073C16Rik
Genomic Map