siRNA Details
| Gene Symbol: 4930502A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040737.1 | 4 | 472 | |||
| GenBank mRNA | AK015674.1 | - | 4 | 472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930502A04Rik| Gene Symbol: 4930502A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040737.1 | 4 | 472 | |||
| GenBank mRNA | AK015674.1 | - | 4 | 472 | ||