siRNA Details
| Gene Symbol: 4930502E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046281.1 | 3 | 670 | |||
| GenBank mRNA | AK015677.1 | - | 3 | 671 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930502E09Rik| Gene Symbol: 4930502E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046281.1 | 3 | 670 | |||
| GenBank mRNA | AK015677.1 | - | 3 | 671 | ||