siRNA Details
| Gene Symbol: 4930502E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046281.1 | 2 | 325 | |||
| GenBank mRNA | AK015677.1 | - | 2 | 326 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715216.1 | - | 326 | Chr. - - [-] on Build | ||

4930502E09Rik
Genomic Map