siRNA Details
| Gene Symbol: 4930546E12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016050.1 | - | 1 | 568 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715646.1 | - | 1 | 567 | Chr. 2 - 115333947 - 115334540 [-] on Build GRCm38 | |

4930546E12Rik
Genomic Map