siRNA Details
| Gene Symbol: 4930525D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040738.1 | 6 | 900 | |||
| GenBank mRNA | AK015892.1 | - | 6 | 901 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF197815.1 | - | 3 | 536 | Chr. 9 - 114341123 - 114355866 [-] on Build GRCm38 | |

4930525D18Rik
Genomic Map