siRNA Details
| Gene Symbol: 4930525D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040738.1 | 5 | 531 | |||
| GenBank mRNA | AK015892.1 | - | 5 | 532 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930525D18Rik| Gene Symbol: 4930525D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040738.1 | 5 | 531 | |||
| GenBank mRNA | AK015892.1 | - | 5 | 532 | ||