siRNA Details
| Gene Symbol: C530007A02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021269.1 | - | 11 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY729319.1 | - | 1 | 59 | Chr. 4 - 132882717 - 132883361 [-] on Build GRCm38 | |

C530007A02Rik
Genomic Map