siRNA Details
| Gene Symbol: C430049B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038184.1 | 4 | 920 | |||
| NR_038185.1 | 4 | 634 | ||||
| GenBank mRNA | AK021262.1 | - | 4 | 635 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU518721.1 | - | 4 | 615 | Chr. X - 53054898 - 53057171 [-] on Build GRCm38 | |

C430049B03Rik
Genomic Map