siRNA Details
Gene Symbol: C430049B03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038184.1 | 4 | 872 | |||
NR_038185.1 | 4 | 586 | ||||
GenBank mRNA | AK021262.1 | - | 4 | 587 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C430049B03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038184.1 | 4 | 872 | |||
NR_038185.1 | 4 | 586 | ||||
GenBank mRNA | AK021262.1 | - | 4 | 587 |