siRNA Details
| Gene Symbol: 5830407E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017905.1 | - | 1 | 1420 | ||
| BB234837.1 | - | 1 | 128 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5830407E08Rik| Gene Symbol: 5830407E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017905.1 | - | 1 | 1420 | ||
| BB234837.1 | - | 1 | 128 | |||