siRNA Details
| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017901.1 | - | 1 | 1685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkce| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017901.1 | - | 1 | 1685 | ||