siRNA Details
Gene Symbol: Arhgap27os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052773.1 | - | 2 | 391 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB661624.1 | - | 2 | 391 | Chr. 11 - 103355701 - 103360327 [+] on Build GRCm38 |
