siRNA Details
| Gene Symbol: Arhgap27os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052773.1 | - | 2 | 243 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB661624.1 | - | 2 | 243 | Chr. 11 - 103355701 - 103360327 [+] on Build GRCm38 | |

Arhgap27os2
Genomic Map