siRNA Details
| Gene Symbol: Adap2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053136.1 | - | 1 | 110 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB605614.2 | - | 1 | 110 | Chr. 11 - 80159176 - 80177629 [-] on Build GRCm38 | |

Adap2os
Genomic Map