siRNA Details
| Gene Symbol: Adap2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053136.1 | - | 1, 2 | 465 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adap2os| Gene Symbol: Adap2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053136.1 | - | 1, 2 | 465 | ||