siRNA Details
| Gene Symbol: Adap2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053136.1 | - | 3 | 689 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB530857.2 | - | 3 | 364 | Chr. 11 - 80158864 - 80177303 [-] on Build GRCm38 | |

Adap2os
Genomic Map