siRNA Details
Gene Symbol: Gm30103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053423.1 | - | 1852 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB539122.2 | - | 1 | 362 | Chr. 5 - 4784753 - 4785411 [-] on Build GRCm38 |
