siRNA Details
Gene Symbol: C330013E15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045701.1 | 1 | 80 | |||
GenBank mRNA | AK021197.1 | - | 1 | 66 | ||
AK083680.1 | - | 1 | 87 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C330013E15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045701.1 | 1 | 80 | |||
GenBank mRNA | AK021197.1 | - | 1 | 66 | ||
AK083680.1 | - | 1 | 87 |