siRNA Details
| Gene Symbol: Ptprtos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040617.1 | 2 | 82 | |||
| GenBank mRNA | AK021192.1 | - | 2 | 83 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprtos| Gene Symbol: Ptprtos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040617.1 | 2 | 82 | |||
| GenBank mRNA | AK021192.1 | - | 2 | 83 | ||