siRNA Details
| Gene Symbol: 1700010B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005813.1 | - | 1 | 978 | ||
| AK133430.1 | - | 1 | 958 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700010B13Rik| Gene Symbol: 1700010B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005813.1 | - | 1 | 978 | ||
| AK133430.1 | - | 1 | 958 | |||