siRNA Details
Gene Symbol: Gm16596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045751.1 | 5 | 1000 | |||
NR_045752.1 | 4 | 950 | ||||
NR_045753.1 | 3 | 892 | ||||
GenBank mRNA | AK140072.1 | - | 4 | 945 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm16596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045751.1 | 5 | 1000 | |||
NR_045752.1 | 4 | 950 | ||||
NR_045753.1 | 3 | 892 | ||||
GenBank mRNA | AK140072.1 | - | 4 | 945 |