siRNA Details
Gene Symbol: Gm16596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045751.1 | 1 | 95 | |||
NR_045752.1 | 1 | 95 | ||||
NR_045753.1 | 1 | 95 | ||||
GenBank mRNA | AK140072.1 | - | 1 | 92 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK618228.1 | - | 1 | 95 | Chr. 12 - 108537344 - 108555618 [-] on Build GRCm38 |
