siRNA Details
| Gene Symbol: Gm16755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131899.1 | - | 2, 3 | 237 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ043460.1 | - | 2, 3 | 237 | Chr. 11 - 120487541 - 120488582 [-] on Build GRCm38 | |

Gm16755
Genomic Map