siRNA Details
Gene Symbol: 5930409G06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020028.1 | - | 1 | 1190 | ||
AK080646.1 | - | 1 | 1108 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5930409G06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020028.1 | - | 1 | 1190 | ||
AK080646.1 | - | 1 | 1108 |