siRNA Details
| Gene Symbol: Ints7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020029.1 | - | 13 | |||
| AK034856.1 | - | 20 | 3876 | |||
| AK052103.1 | - | 18 | 3603 | |||
| BC054120.1 | - | 20 | 3863 | 6506658 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ints7| Gene Symbol: Ints7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020029.1 | - | 13 | |||
| AK034856.1 | - | 20 | 3876 | |||
| AK052103.1 | - | 18 | 3603 | |||
| BC054120.1 | - | 20 | 3863 | 6506658 | ||