siRNA Details
| Gene Symbol: Gm20139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038021.1 | 4 | 3507 | |||
| NR_038022.1 | 2 | 3239 | ||||
| GenBank mRNA | AK044491.1 | - | 4 | 3508 | ||
| AK137933.1 | - | 1 | 2884 | |||
| AK169479.1 | - | 2 | 3240 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20139| Gene Symbol: Gm20139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038021.1 | 4 | 3507 | |||
| NR_038022.1 | 2 | 3239 | ||||
| GenBank mRNA | AK044491.1 | - | 4 | 3508 | ||
| AK137933.1 | - | 1 | 2884 | |||
| AK169479.1 | - | 2 | 3240 | |||