siRNA Details
| Gene Symbol: Gm20139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038021.1 | 4 | 801 | |||
| NR_038022.1 | 2 | 533 | ||||
| GenBank mRNA | AK044491.1 | - | 4 | 802 | ||
| AK137933.1 | - | 1 | 179 | |||
| AK169479.1 | - | 2 | 534 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB638474.1 | - | 2 | 533 | Chr. 10 - 19167238 - 19202601 [+] on Build GRCm38 | |

Gm20139
Genomic Map