siRNA Details
| Gene Symbol: Stk38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020024.1 | - | 1 | 704 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA526734.1 | - | 1 | 375 | Chr. 17 - 29006991 - 29007539 [-] on Build GRCm38 | |

Stk38
Genomic Map