siRNA Details
| Gene Symbol: Jakmip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051838.1 | - | 1 | 741 | ||
| AK086707.1 | - | 1 | 841 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jakmip2| Gene Symbol: Jakmip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051838.1 | - | 1 | 741 | ||
| AK086707.1 | - | 1 | 841 | |||