siRNA Details
| Gene Symbol: B130055M24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041042.1 | - | 1 | 864 | ||
| AK045290.1 | - | 1 | 856 | |||
| AK050836.1 | - | 1 | 860 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB221467.2 | - | 1 | 411 | Chr. 7 - 127449089 - 127457185 [+] on Build GRCm38 | |

B130055M24Rik
Genomic Map