siRNA Details
Gene Symbol: B130055M24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041042.1 | - | 1 | 464 | ||
AK045290.1 | - | 1 | 456 | |||
AK050836.1 | - | 1 | 460 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B130055M24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041042.1 | - | 1 | 464 | ||
AK045290.1 | - | 1 | 456 | |||
AK050836.1 | - | 1 | 460 |