siRNA Details
| Gene Symbol: 1700018P22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006100.1 | - | 1 | 110 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706212.1 | - | 1 | 110 | Chr. 9 - 8134513 - 8135844 [+] on Build GRCm38 | |

1700018P22Rik
Genomic Map