siRNA Details
| Gene Symbol: 3300002P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI430424.1 | - | 1 | 143 | ||
| AK014377.1 | - | 1 | 520 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3300002P09Rik| Gene Symbol: 3300002P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI430424.1 | - | 1 | 143 | ||
| AK014377.1 | - | 1 | 520 | |||