siRNA Details
| Gene Symbol: Gm29514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158929.1 | - | 2189 | |||
| Gene Symbol: 4933405D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046036.1 | 6 | 740 | |||
| GenBank mRNA | AK045097.1 | - | 6 | 741 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB294941.2 | - | 5 | 678 | Chr. 3 - 122882592 - 122919780 [-] on Build GRCm38 | |

Gm29514
Genomic Map