siRNA Details
| Gene Symbol: Sesn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033655.1 | - | 1202 | |||
| BC003348.1 | - | 738 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sesn3| Gene Symbol: Sesn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033655.1 | - | 1202 | |||
| BC003348.1 | - | 738 | ||||