siRNA Details
| Gene Symbol: 1110002E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003290.1 | - | 812 | |||
| AK009907.1 | - | 283 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110002E22Rik| Gene Symbol: 1110002E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003290.1 | - | 812 | |||
| AK009907.1 | - | 283 | ||||