siRNA Details
| Gene Symbol: 4930441J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040501.1 | 5 | 408 | |||
| GenBank mRNA | AK015352.1 | - | 5 | 408 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714907.1 | - | 5 | 405 | Chr. 2 - 74298938 - 74307714 [-] on Build GRCm38 | |

4930441J16Rik
Genomic Map