siRNA Details
| Gene Symbol: 4930448O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015424.1 | - | 1 | 438 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714972.1 | - | 1 | 438 | Chr. 13 - 97980802 - 97981452 [+] on Build GRCm38 | |

4930448O17Rik
Genomic Map