siRNA Details
| Gene Symbol: Gm13986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048775.1 | - | 1 | 292 | ||
| AK054543.1 | - | 1 | 184 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB666966.1 | - | 1 | 180 | Chr. 2 - 118071764 - 118111091 [-] on Build GRCm38 | |

Gm13986
Genomic Map