siRNA Details
| Gene Symbol: A630023P12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102290.1 | 1 | 73 | |||
| GenBank mRNA | AK088579.1 | - | 1 | 74 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK040026.1 | - | 52 | Chr. - - [-] on Build | ||

A630023P12Rik
Genomic Map