siRNA Details
| Gene Symbol: Gm6260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040405.1 | 2 | 235 | |||
| GenBank mRNA | AK043613.1 | - | 2 | 237 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB641225.1 | - | 2 | 237 | Chr. 3 - 143671134 - 143712143 [-] on Build GRCm38 | |

Gm6260
Genomic Map