siRNA Details
| Gene Symbol: Gm6260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040405.1 | 3 | 686 | |||
| GenBank mRNA | AK043613.1 | - | 3 | 688 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6260| Gene Symbol: Gm6260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040405.1 | 3 | 686 | |||
| GenBank mRNA | AK043613.1 | - | 3 | 688 | ||