siRNA Details
| Gene Symbol: Gm5833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040304.1 | 3 | 860 | |||
| GenBank mRNA | AK154631.1 | - | 3 | 862 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5833| Gene Symbol: Gm5833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040304.1 | 3 | 860 | |||
| GenBank mRNA | AK154631.1 | - | 3 | 862 | ||