siRNA Details
| Gene Symbol: A230070E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045897.1 | 1 | 362 | |||
| GenBank mRNA | AK048964.1 | - | 1 | 363 | ||
| AK162870.1 | - | 1 | 363 | |||
| AK164995.1 | - | 1 | 430 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230070E04Rik| Gene Symbol: A230070E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045897.1 | 1 | 362 | |||
| GenBank mRNA | AK048964.1 | - | 1 | 363 | ||
| AK162870.1 | - | 1 | 363 | |||
| AK164995.1 | - | 1 | 430 | |||