siRNA Details
| Gene Symbol: Gm15201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136312.1 | - | 2 | 921 | ||
| BB065474.2 | - | 1 | 388 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15201| Gene Symbol: Gm15201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136312.1 | - | 2 | 921 | ||
| BB065474.2 | - | 1 | 388 | |||