siRNA Details
| Gene Symbol: Gm15201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136312.1 | - | 2 | 578 | ||
| BB065474.2 | - | 1 | 45 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB624209.1 | - | 2 | 574 | Chr. X - 162735639 - 162736611 [-] on Build GRCm38 | |

Gm15201
Genomic Map