siRNA Details
| Gene Symbol: 4930579P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016333.1 | - | 3 | 594 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715974.1 | - | 594 | Chr. - - [-] on Build | ||

4930579P08Rik
Genomic Map