siRNA Details
| Gene Symbol: 4933435F18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017068.1 | - | 2 | 431 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716748.1 | - | 2 | 431 | Chr. 4 - 124331404 - 124332467 [-] on Build GRCm38 | |

4933435F18Rik
Genomic Map