siRNA Details
| Gene Symbol: 4933435G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017069.1 | - | 1 | 1695 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN840989.1 | - | 323 | Chr. - - [-] on Build | ||

4933435G04Rik
Genomic Map